Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTGCCTATGAGCAGCTGGGCTAC[C/T]GTGCCTTTGGGACCCCAGGAAAGCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604437 | |||||||||||||||||||||||
Literature Links: |
SLC38A3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC38A3 - solute carrier family 38 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006841.5 | 557 | Missense Mutation | CGT,TGT | R,C 135 | NP_006832.1 | |
XM_006712954.2 | 557 | Missense Mutation | CGT,TGT | R,C 135 | XP_006713017.1 |