Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTTTGCTGAGGTGCTGCGGCTG[C/G]CGAAGCAGCAGCTGAGGAAGCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GTPBP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTPBP8 - GTP binding protein 8 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014170.2 | 165 | Missense Mutation | CCG,GCG | P,A 40 | NP_054889.2 | |
NM_138485.1 | 165 | Missense Mutation | CCG,GCG | P,A 40 | NP_612494.1 |