Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGGAGAAGTACTCCGGGCTCAC[C/T]TGAAAATAGTTACGAATGTCTTTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608298 | |||||||||||||||||||||||
Literature Links: |
CCDC80 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC80 - coiled-coil domain containing 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199511.2 | 2805 | Silent Mutation | CAA,CAG | Q,Q 858 | NP_955805.1 | |
NM_199512.2 | 2805 | Silent Mutation | CAA,CAG | Q,Q 858 | NP_955806.1 |
LINC01279 - long intergenic non-protein coding RNA 1279 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |