Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAGCTCCTCCGGTACCGGCTGG[C/G]CGAGGAGGGCCCCGCCGACGTCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602988 | ||||||||||||||||||||
Literature Links: |
PCDH7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDH7 - protocadherin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173523.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | NP_001166994.1 | |
NM_002589.2 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | NP_002580.2 | |
NM_032456.2 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | NP_115832.1 | |
NM_032457.3 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | NP_115833.2 | |
XM_005248163.3 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_005248220.1 | |
XM_005248164.3 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_005248221.1 | |
XM_005248166.3 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_005248223.1 | |
XM_011513842.2 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_011512144.1 | |
XM_011513843.2 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_011512145.1 | |
XM_011513844.2 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_011512146.1 | |
XM_017008259.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863748.1 | |
XM_017008260.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863749.1 | |
XM_017008261.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863750.1 | |
XM_017008262.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863751.1 | |
XM_017008263.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863752.1 | |
XM_017008264.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863753.1 | |
XM_017008265.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863754.1 | |
XM_017008266.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863755.1 | |
XM_017008267.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863756.1 | |
XM_017008268.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863757.1 | |
XM_017008269.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863758.1 | |
XM_017008270.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863759.1 | |
XM_017008271.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863760.1 | |
XM_017008272.1 | 1160 | Missense Mutation | GCC,GGC | A,G 39 | XP_016863761.1 |