Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCTTCTTTAGAAAGCACATCTT[G/T]TAGGATGGAGATAGTAGAAATGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607590 MIM: 123835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBS7 - Bardet-Biedl syndrome 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018190.3 | 1913 | Missense Mutation | AAA,CAA | K,Q 577 | NP_060660.2 | |
NM_176824.2 | 1913 | Missense Mutation | AAA,CAA | K,Q 577 | NP_789794.1 | |
XM_005263106.3 | 1913 | Missense Mutation | AAA,CAA | K,Q 578 | XP_005263163.1 | |
XM_011532079.2 | 1913 | Missense Mutation | AAA,CAA | K,Q 593 | XP_011530381.1 | |
XM_011532080.2 | 1913 | Missense Mutation | AAA,CAA | K,Q 592 | XP_011530382.1 | |
XM_011532081.2 | 1913 | Missense Mutation | AAA,CAA | K,Q 538 | XP_011530383.1 | |
XM_017008357.1 | 1913 | Missense Mutation | AAA,CAA | K,Q 522 | XP_016863846.1 | |
XM_017008358.1 | 1913 | Missense Mutation | AAA,CAA | K,Q 523 | XP_016863847.1 |
CCNA2 - cyclin A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |