Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAACAGCTGCTCCTCCTCAGCC[A/G]TGTCCCCGGGGCCCTCCCCGAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 252800 MIM: 610130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDUA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IDUA - iduronidase, alpha-L- | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000203.4 | 2176 | Intron | NP_000194.2 | |||
XM_011513461.2 | 2176 | Intron | XP_011511763.1 | |||
XM_017008163.1 | 2176 | Intron | XP_016863652.1 |
SLC26A1 - solute carrier family 26 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022042.3 | 2176 | Missense Mutation | ACG,ATG | T,M 670 | NP_071325.2 | |
NM_134425.2 | 2176 | Intron | NP_602297.1 | |||
NM_213613.3 | 2176 | Missense Mutation | ACG,ATG | T,M 670 | NP_998778.1 |