Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATGCCGATGATGATGGTGGGAG[A/C]TCCGTGCAAAGTGAAGCTGAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611614 | ||||||||||||||||||||
Literature Links: |
UTP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UTP3 - UTP3, small subunit processome component homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020368.2 | 538 | Missense Mutation | AGA,AGC | R,S 113 | NP_065101.1 |