Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAAATTTGCCTCAAGATGGAAA[C/T]CCTTTGCCTCAGGGCATCCTTTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 131243 | ||||||||||||||||||||
Literature Links: |
EDNRA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EDNRA - endothelin receptor type A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166055.1 | 538 | Missense Mutation | ACC,ATC | T,I 3 | NP_001159527.1 | |
NM_001256283.1 | 538 | Intron | NP_001243212.1 | |||
NM_001957.3 | 538 | Missense Mutation | ACC,ATC | T,I 3 | NP_001948.1 |