Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGAGTACAAAAAGCAGGAGTAC[A/G]TGGTGGTCGAGGCGGAGGAAGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENPEP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENPEP - glutamyl aminopeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001977.3 | 847 | Missense Mutation | ATG,GTG | M,V 169 | NP_001968.3 | |
XM_017007877.1 | 847 | Intron | XP_016863366.1 |