Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCACCGTGGTGGCTCTGCTCACC[A/G]TGACGGACGCAGATTCTCCCGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAT4 - FAT atypical cadherin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291285.1 | 1144 | Missense Mutation | ATG,GTG | M,V 382 | NP_001278214.1 | |
NM_001291303.1 | 1144 | Missense Mutation | ATG,GTG | M,V 382 | NP_001278232.1 | |
NM_024582.4 | 1144 | Missense Mutation | ATG,GTG | M,V 382 | NP_078858.4 | |
XM_011532236.2 | 1144 | Missense Mutation | ATG,GTG | M,V 382 | XP_011530538.1 | |
XM_011532237.2 | 1144 | Intron | XP_011530539.1 |