Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTAAAAGGCTGCTGTACCTCTG[A/G]CTGGCACTTCTGGCTTGCTGTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNI - cyclin I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006835.2 | 1268 | Missense Mutation | GCC,GTC | A,V 300 | NP_006826.1 | |
XM_011531555.1 | 1268 | Missense Mutation | GCC,GTC | A,V 228 | XP_011529857.1 |
SEPT11 - septin 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |