Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGATCTGTGATGCTCATCTGTCAC[C/T]GTGTAGTACTGAGCTAAAACCTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LYAR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LYAR - Ly1 antibody reactive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145725.1 | 1200 | Silent Mutation | ACA,ACG | T,T 342 | NP_001139197.1 | |
NM_017816.2 | 1200 | Silent Mutation | ACA,ACG | T,T 342 | NP_060286.1 | |
XM_011513505.1 | 1200 | Silent Mutation | ACA,ACG | T,T 342 | XP_011511807.1 | |
XM_011513506.2 | 1200 | Silent Mutation | ACA,ACG | T,T 342 | XP_011511808.1 |