Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCCGAGAACGAGCGGTGGAGAT[G/T]GATGGGGAGCGCATCAAGGTGAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605950 | ||||||||||||||||||||
Literature Links: |
RAB33B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB33B - RAB33B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031296.2 | 716 | Missense Mutation | ATG,ATT | M,I 77 | NP_112586.1 | |
XM_011532299.1 | 716 | Missense Mutation | ATG,ATT | M,I 125 | XP_011530601.1 |