Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATCCACTGATTCTAGACAACAAA[C/G]TGGATCACCTAAAAAGAGTGCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN2AIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN2AIP - CDKN2A interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317343.1 | 973 | UTR 3 | NP_001304272.1 | |||
NM_017632.3 | 973 | Missense Mutation | ACT,AGT | T,S 266 | NP_060102.1 |