Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGAAGTTTATATTTCTCCAAA[T/A]CAATTTCTGGAAAAAACGTGTCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126060 | ||||||||||||||||||||
Literature Links: |
DHFR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHFR - dihydrofolate reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000791.3 | 950 | Missense Mutation | GAT,GTT | D,V 153 | NP_000782.1 | |
NM_001290354.1 | 950 | Missense Mutation | GAT,GTT | D,V 101 | NP_001277283.1 | |
NM_001290357.1 | 950 | Intron | NP_001277286.1 |