Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCCCATGGTGCCAGTCATCACA[C/T]TGGGCATGCTCATAGGCATGGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607265 | ||||||||||||||||||||
Literature Links: |
CLINT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLINT1 - clathrin interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195555.1 | 1869 | Missense Mutation | AAT,AGT | N,S 573 | NP_001182484.1 | |
NM_001195556.1 | 1869 | Missense Mutation | AAT,AGT | N,S 555 | NP_001182485.1 | |
NM_014666.3 | 1869 | Missense Mutation | AAT,AGT | N,S 555 | NP_055481.1 | |
XM_011534701.2 | 1869 | UTR 3 | XP_011533003.1 | |||
XM_017010087.1 | 1869 | UTR 3 | XP_016865576.1 | |||
XM_017010088.1 | 1869 | Intron | XP_016865577.1 |