Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTTCAAGAGAACTATGATCGTC[C/T]ATCAATGGAGAGATTTTCGTATTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIN1 - gypsy retrotransposon integrase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317954.1 | 1379 | Missense Mutation | ATA,ATG | I,M 350 | NP_001304883.1 | |
NM_017676.2 | 1379 | Missense Mutation | ATA,ATG | I,M 497 | NP_060146.2 | |
XM_005272028.3 | 1379 | Missense Mutation | ATA,ATG | I,M 350 | XP_005272085.1 | |
XM_017009599.1 | 1379 | Missense Mutation | ATA,ATG | I,M 497 | XP_016865088.1 |