Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGTCCTTCAGGGAAGATGGGA[A/G]GCGAGGAGAAGCCGGTGAGCAAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TARS - threonyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258437.1 | 354 | Missense Mutation | AGC,GGC | S,G 15 | NP_001245366.1 | |
NM_001258438.1 | 354 | Missense Mutation | AGC,GGC | S,G 15 | NP_001245367.1 | |
NM_152295.4 | 354 | Missense Mutation | AGC,GGC | S,G 15 | NP_689508.3 |