Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACGTCTGCAGGCCAGTCCTTTC[C/T]GGCTAGTGCCTAATGTCAGAATCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615762 MIM: 613377 | ||||||||||||||||||||
Literature Links: |
GRXCR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRXCR2 - glutaredoxin and cysteine rich domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3RF2 - SH3 domain containing ring finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152550.3 | 577 | Missense Mutation | CGG,TGG | R,W 115 | NP_689763.3 | |
XM_005268380.2 | 577 | Missense Mutation | CGG,TGG | R,W 115 | XP_005268437.1 | |
XM_006714757.1 | 577 | Missense Mutation | CGG,TGG | R,W 115 | XP_006714820.1 | |
XM_011537567.2 | 577 | Missense Mutation | CGG,TGG | R,W 115 | XP_011535869.1 | |
XM_011537568.2 | 577 | Missense Mutation | CGG,TGG | R,W 115 | XP_011535870.1 | |
XM_011537569.2 | 577 | Missense Mutation | CGG,TGG | R,W 115 | XP_011535871.1 | |
XM_017009136.1 | 577 | Missense Mutation | CGG,TGG | R,W 115 | XP_016864625.1 |