Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGAGCAAGGCCGGCTGGCAGAA[C/T]TCCATCCGCCACAACCTGTCGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601093 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXI1 - forkhead box I1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012188.4 | 552 | Silent Mutation | AAC,AAT | N,N 169 | NP_036320.2 | |
NM_144769.2 | 552 | Silent Mutation | AAC,AAT | N,N 169 | NP_658982.1 |