Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACCTCAGCCCTGAGGTGCCCAG[A/C]AATGCCACCTTTCACAGCTGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609383 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724404 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102724404 - uncharacterized LOC102724404 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NIPAL4 - NIPA like domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099287.1 | 419 | Silent Mutation | AGA,AGC | R,S 101 | NP_001092757.1 | |
NM_001172292.1 | 419 | Silent Mutation | AGA,AGC | R,S 101 | NP_001165763.1 | |
XM_011534552.1 | 419 | UTR 5 | XP_011532854.1 |