Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACCAAACTGCCCACTTCGTTG[C/T]TTGCTGAGACTCTCGGGCCAGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 139150 | ||||||||||||||||||||
Literature Links: |
RASA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RASA1 - RAS p21 protein activator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002890.2 | 1021 | Missense Mutation | CTT,TTT | L,F 124 | NP_002881.1 | |
NM_022650.2 | 1021 | Intron | NP_072179.1 | |||
XM_011543525.1 | 1021 | Missense Mutation | CTT,TTT | L,F 124 | XP_011541827.1 | |
XM_011543527.2 | 1021 | Missense Mutation | CTT,TTT | L,F 124 | XP_011541829.1 |