Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTATGAGGCACATCTTGATTCT[C/T]ATTATGAGATAATAAAATGATTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRPC7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRPC7 - transient receptor potential cation channel subfamily C member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167576.1 | 2043 | Missense Mutation | ATA,ATG | I,M 607 | NP_001161048.1 | |
NM_001167577.1 | 2043 | Missense Mutation | ATA,ATG | I,M 662 | NP_001161049.1 | |
NM_020389.2 | 2043 | Missense Mutation | ATA,ATG | I,M 723 | NP_065122.1 |
TRPC7-AS2 - TRPC7 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |