Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGGGGACACTCCCCACATGTT[A/C]CTTGACATCACACAGAAGGGCAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603335 | ||||||||||||||||||||
Literature Links: |
DNAH5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAH5 - dynein axonemal heavy chain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017009180.1 | Intron | XP_016864669.1 | ||||
XM_017009181.1 | Intron | XP_016864670.1 | ||||
XM_017009182.1 | Intron | XP_016864671.1 | ||||
XM_017009183.1 | Intron | XP_016864672.1 | ||||
XM_017009184.1 | Intron | XP_016864673.1 | ||||
XM_017009187.1 | Intron | XP_016864676.1 |