Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACAGCCAACTGCCACTTCAAC[A/G]AAAGAGAAACAGCCATGCTCAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BDP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BDP1 - B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018429.2 | 641 | Silent Mutation | ACA,ACG | T,T 133 | NP_060899.2 | |
XM_011543511.2 | 641 | Silent Mutation | ACA,ACG | T,T 133 | XP_011541813.1 | |
XM_011543512.2 | 641 | Silent Mutation | ACA,ACG | T,T 133 | XP_011541814.1 | |
XM_017009630.1 | 641 | Silent Mutation | ACA,ACG | T,T 133 | XP_016865119.1 | |
XM_017009631.1 | 641 | Silent Mutation | ACA,ACG | T,T 133 | XP_016865120.1 | |
XM_017009632.1 | 641 | Silent Mutation | ACA,ACG | T,T 133 | XP_016865121.1 | |
XM_017009633.1 | 641 | Silent Mutation | ACA,ACG | T,T 133 | XP_016865122.1 | |
XM_017009634.1 | 641 | Silent Mutation | ACA,ACG | T,T 133 | XP_016865123.1 |