Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACCGTCCTCGCTCTGGTTGTA[G/A]TAGCCGCGCAGGTTCCGCAGGCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142840 | ||||||||||||||||||||
Literature Links: |
HLA-C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HLA-C - major histocompatibility complex, class I, C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243042.1 | 389 | Silent Mutation | TAC,TAT | Y,Y 108 | NP_001229971.1 | |
NM_002117.5 | 389 | Silent Mutation | TAC,TAT | Y,Y 108 | NP_002108.4 |