Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCACCCATGGCAGAAGGAGGAG[A/G]GCAGAATCATCACGAAGGTGAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VEGFA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VEGFA - vascular endothelial growth factor A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025366.2 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001020537.2 | |
NM_001025367.2 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001020538.2 | |
NM_001025368.2 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001020539.2 | |
NM_001025369.2 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001020540.2 | |
NM_001025370.2 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001020541.2 | |
NM_001033756.2 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001028928.1 | |
NM_001171622.1 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001165093.1 | |
NM_001171623.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165094.1 | |
NM_001171624.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165095.1 | |
NM_001171625.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165096.1 | |
NM_001171626.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165097.1 | |
NM_001171627.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165098.1 | |
NM_001171628.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165099.1 | |
NM_001171629.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165100.1 | |
NM_001171630.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001165101.1 | |
NM_001204384.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001191313.1 | |
NM_001204385.1 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_001191314.1 | |
NM_001287044.1 | 1139 | Missense Mutation | GAG,GGG | E,G 6 | NP_001273973.1 | |
NM_001317010.1 | 1139 | Missense Mutation | GAG,GGG | E,G 34 | NP_001303939.1 | |
NM_003376.5 | 1139 | Missense Mutation | GAG,GGG | E,G 214 | NP_003367.4 |