Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTTTTTCTTTTTCAGCATCTA[C/T]TACAAGCCTTAGTGGGTCTGATAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616189 | ||||||||||||||||||||
Literature Links: |
CMTR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CMTR1 - cap methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015050.2 | 393 | Missense Mutation | ACT,ATT | T,I 47 | NP_055865.1 | |
XM_005248955.3 | 393 | Missense Mutation | ACT,ATT | T,I 47 | XP_005249012.1 |