Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCATCTCCCTCACCCCCAGTGT[C/G]GCTTCAGCTATGCACGGGGACAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL14 - mitochondrial ribosomal protein L14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM63B - transmembrane protein 63B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318792.1 | 487 | Silent Mutation | GTC,GTG | V,V 94 | NP_001305721.1 | |
NM_018426.2 | 487 | Silent Mutation | GTC,GTG | V,V 94 | NP_060896.1 | |
XM_005249211.3 | 487 | Silent Mutation | GTC,GTG | V,V 103 | XP_005249268.1 | |
XM_005249213.4 | 487 | Silent Mutation | GTC,GTG | V,V 94 | XP_005249270.1 | |
XM_005249214.4 | 487 | Silent Mutation | GTC,GTG | V,V 90 | XP_005249271.1 | |
XM_005249215.4 | 487 | Silent Mutation | GTC,GTG | V,V 81 | XP_005249272.1 | |
XM_005249217.1 | 487 | Silent Mutation | GTC,GTG | V,V 94 | XP_005249274.1 | |
XM_006715135.1 | 487 | Silent Mutation | GTC,GTG | V,V 94 | XP_006715198.1 | |
XM_017010999.1 | 487 | Silent Mutation | GTC,GTG | V,V 103 | XP_016866488.1 | |
XM_017011000.1 | 487 | Silent Mutation | GTC,GTG | V,V 94 | XP_016866489.1 | |
XM_017011001.1 | 487 | Silent Mutation | GTC,GTG | V,V 81 | XP_016866490.1 | |
XM_017011002.1 | 487 | Silent Mutation | GTC,GTG | V,V 81 | XP_016866491.1 |