Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACTCTCACAACTGCACCCTTGC[C/T]GGGCACCACTCCACTGTATCCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602017 MIM: 600075 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMB1 - proteasome subunit beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBP - TATA-box binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172085.1 | 475 | Missense Mutation | CCG,CTG | P,L 113 | NP_001165556.1 | |
NM_003194.4 | 475 | Missense Mutation | CCG,CTG | P,L 133 | NP_003185.1 |