Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGCGACAGCGGCTTGGAAGTGC[C/T]CTACCAGAGTTGCGGTAAGCCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605241 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LY86 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LY86 - lymphocyte antigen 86 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004271.3 | 218 | Missense Mutation | CCC,CTC | P,L 41 | NP_004262.1 | |
XM_017011505.1 | 218 | Missense Mutation | CCC,CTC | P,L 41 | XP_016866994.1 |
LY86-AS1 - LY86 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |