Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATCATCTGAACTATCCTCAGA[C/T]TCACTTTCTAAAAGTAATATAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125264 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEK - DEK proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134709.1 | 997 | Silent Mutation | GAA,GAG | E,E 268 | NP_001128181.1 | |
NM_003472.3 | 997 | Silent Mutation | GAA,GAG | E,E 302 | NP_003463.1 | |
XM_017011273.1 | 997 | Silent Mutation | GAA,GAG | E,E 302 | XP_016866762.1 |