Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTCAGCCTTCTGCATCTGGATC[C/T]CTTGTTGGAAGAGAAAATGAGGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602842 | ||||||||||||||||||||
Literature Links: |
GMNN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMNN - geminin, DNA replication inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251989.1 | 239 | Silent Mutation | TCC,TCT | S,S 36 | NP_001238918.1 | |
NM_001251990.1 | 239 | Silent Mutation | TCC,TCT | S,S 36 | NP_001238919.1 | |
NM_001251991.1 | 239 | Silent Mutation | TCC,TCT | S,S 36 | NP_001238920.1 | |
NM_015895.4 | 239 | Silent Mutation | TCC,TCT | S,S 36 | NP_056979.1 | |
XM_005249159.1 | 239 | Silent Mutation | TCC,TCT | S,S 36 | XP_005249216.1 | |
XM_011514651.2 | 239 | Silent Mutation | TCC,TCT | S,S 30 | XP_011512953.1 |