Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGACCTTGCAGAGCAGCAAGAGGC[C/T]CTCTCCAGAAAACATCGGCAGATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609091 | ||||||||||||||||||||
Literature Links: |
FBXO9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO9 - F-box protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012347.4 | 398 | Missense Mutation | CCT,TCT | P,S 74 | NP_036479.1 | |
NM_033480.2 | 398 | Missense Mutation | CCT,TCT | P,S 64 | NP_258441.1 | |
NM_033481.3 | 398 | Missense Mutation | CCT,TCT | P,S 30 | NP_258442.2 | |
XM_005248995.4 | 398 | Missense Mutation | CCT,TCT | P,S 30 | XP_005249052.1 | |
XM_005248996.4 | 398 | Missense Mutation | CCT,TCT | P,S 30 | XP_005249053.1 |
LOC105375092 - uncharacterized LOC105375092 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |