Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTATTACCACGTCCCAGTGAAGC[A/G]CTCTGAGAAGGCCGTGGATGCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf57 - chromosome 7 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100159.2 | 148 | Missense Mutation | CAC,CGC | H,R 28 | NP_001093629.1 | |
NM_001267865.1 | 148 | UTR 5 | NP_001254794.1 | |||
NM_001267866.1 | 148 | UTR 5 | NP_001254795.1 | |||
XM_006715649.3 | 148 | Missense Mutation | CAC,CGC | H,R 28 | XP_006715712.1 | |
XM_006715650.2 | 148 | UTR 5 | XP_006715713.1 | |||
XM_011515121.1 | 148 | Missense Mutation | CAC,CGC | H,R 28 | XP_011513423.1 | |
XM_011515122.2 | 148 | Missense Mutation | CAC,CGC | H,R 28 | XP_011513424.1 | |
XM_011515123.2 | 148 | Missense Mutation | CAC,CGC | H,R 28 | XP_011513425.1 |
SUN3 - Sad1 and UNC84 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |