Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAGCTCAGGATCTCGGGTCTCC[A/G]CCGGGACATCTCGTCTATGTCACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611345 | ||||||||||||||||||||
Literature Links: |
INTS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INTS1 - integrator complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080453.2 | 6327 | Silent Mutation | NP_001073922.2 | |||
XM_011515260.1 | 6327 | Missense Mutation | XP_011513562.1 | |||
XM_011515262.2 | 6327 | Intron | XP_011513564.1 | |||
XM_017011959.1 | 6327 | Missense Mutation | XP_016867448.1 | |||
XM_017011960.1 | 6327 | Missense Mutation | XP_016867449.1 |