Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGACATGAGATGGTAGACAAA[A/G]CCCCAGGTAGGTGACAGTTAATACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603974 | ||||||||||||||||||||
Literature Links: |
ZNF92 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF92 - zinc finger protein 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287532.1 | 419 | Intron | NP_001274461.1 | |||
NM_001287533.1 | 419 | UTR 5 | NP_001274462.1 | |||
NM_001287534.1 | 419 | Intron | NP_001274463.1 | |||
NM_007139.3 | 419 | Missense Mutation | ACC,GCC | T,A 5 | NP_009070.2 | |
NM_152626.3 | 419 | Missense Mutation | ACC,GCC | T,A 74 | NP_689839.1 |