Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCAGTGAGGGGCTGCGGCCCAC[C/T]CAGGCGGGGAGCCTCTCAGGCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616861 MIM: 614469 MIM: 602933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6875 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6875 - microRNA 6875 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A9 - solute carrier family 12 member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRRT - serrate, RNA effector molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIP6 - thyroid hormone receptor interactor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003302.2 | 721 | Missense Mutation | CCC,CTC | P,L 184 | NP_003293.2 |