Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGCAAGCGCCACAGAAGCCTC[A/G]TCTCCTGGAGGAAAATGGTGAGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611272 | ||||||||||||||||||||
Literature Links: |
ZKSCAN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZKSCAN5 - zinc finger with KRAB and SCAN domains 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318082.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | NP_001305011.1 | |
NM_001318083.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | NP_001305012.1 | |
NM_001318084.1 | 666 | UTR 5 | NP_001305013.1 | |||
NM_014569.3 | 666 | Missense Mutation | CAT,CGT | H,R 179 | NP_055384.1 | |
NM_145102.3 | 666 | Missense Mutation | CAT,CGT | H,R 179 | NP_659570.1 | |
XM_011515999.2 | 666 | Missense Mutation | CAT,CGT | H,R 30 | XP_011514301.1 | |
XM_017011918.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | XP_016867407.1 | |
XM_017011919.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | XP_016867408.1 | |
XM_017011920.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | XP_016867409.1 | |
XM_017011921.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | XP_016867410.1 | |
XM_017011922.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | XP_016867411.1 | |
XM_017011923.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | XP_016867412.1 | |
XM_017011924.1 | 666 | Missense Mutation | CAT,CGT | H,R 179 | XP_016867413.1 | |
XM_017011925.1 | 666 | Missense Mutation | CAT,CGT | H,R 30 | XP_016867414.1 | |
XM_017011926.1 | 666 | UTR 5 | XP_016867415.1 | |||
XM_017011927.1 | 666 | UTR 5 | XP_016867416.1 |