Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGGAGGCCCCTCTGGTATTAG[A/T]GAAGTAAAGAAGGTGGTGATGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERPUD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HERPUD2 - HERPUD family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022373.4 | 1652 | Silent Mutation | TCA,TCT | S,S 395 | NP_071768.3 | |
XM_006715765.1 | 1652 | Silent Mutation | TCA,TCT | S,S 331 | XP_006715828.1 | |
XM_011515473.1 | 1652 | Silent Mutation | TCA,TCT | S,S 331 | XP_011513775.1 | |
XM_017012506.1 | 1652 | Silent Mutation | TCA,TCT | S,S 395 | XP_016867995.1 |