Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCAGAGGTACTCCACGCAGATC[C/T]ACAGCTTCCGAGAGGGCACCTAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608606 MIM: 610989 MIM: 614781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHLHA15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BHLHA15 - basic helix-loop-helix family member a15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177455.3 | 603 | Missense Mutation | CAC,TAC | H,Y 183 | NP_803238.1 |
LMTK2 - lemur tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TECPR1 - tectonin beta-propeller repeat containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |