Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCAGTGTCCGGTGCATGAGGAA[A/G]GTGTCCTCTGAAGGGCGGGGCCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEPCE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEPCE - methylphosphate capping enzyme | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R35 - protein phosphatase 1 regulatory subunit 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145030.2 | 567 | Silent Mutation | ACC,ACT | T,T 241 | NP_659467.1 | |
XM_011515914.1 | 567 | Silent Mutation | ACC,ACT | T,T 171 | XP_011514216.1 |
ZCWPW1 - zinc finger CW-type and PWWP domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |