Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCCACTAATGGCTCCCATAGT[C/T]TCCATTCCCTCATTTTTCTTGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601677 | ||||||||||||||||||||
Literature Links: |
IQUB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQUB - IQ motif and ubiquitin domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA5 - NADH:ubiquinone oxidoreductase subunit A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282419.2 | 596 | Missense Mutation | AAA,AGA | K,R 83 | NP_001269348.1 | |
NM_001282420.2 | 596 | Missense Mutation | AAA,AGA | K,R 98 | NP_001269349.1 | |
NM_001282421.2 | 596 | Missense Mutation | AAA,AGA | K,R 95 | NP_001269350.1 | |
NM_001282422.2 | 596 | Missense Mutation | AAA,AGA | K,R 59 | NP_001269351.1 | |
NM_001291304.1 | 596 | Missense Mutation | AAA,AGA | K,R 164 | NP_001278233.1 | |
NM_005000.4 | 596 | Missense Mutation | AAA,AGA | K,R 98 | NP_004991.1 |