Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAGTCTGTAAGTTGTTTGTGTG[C/T]CACTGTGACGGCTCCCACGGTTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610032 | ||||||||||||||||||||
Literature Links: |
TNPO3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNPO3 - transportin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191028.2 | 3074 | Missense Mutation | ACA,GCA | T,A 827 | NP_001177957.2 | |
NM_012470.3 | 3074 | Missense Mutation | ACA,GCA | T,A 891 | NP_036602.1 | |
XM_011515989.2 | 3074 | Missense Mutation | ACA,GCA | T,A 825 | XP_011514291.1 |