Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCTAAAGAAAAAGGCCATGGAT[A/G]TAGAAATTCAGGACACACCTGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608498 | ||||||||||||||||||||
Literature Links: |
C7orf72 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C7orf72 - chromosome 7 open reading frame 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161834.2 | 232 | Missense Mutation | ATA,GTA | I,V 3 | NP_001155306.2 | |
XM_011515052.1 | 232 | Missense Mutation | ATA,GTA | I,V 3 | XP_011513354.1 | |
XM_011515053.2 | 232 | Missense Mutation | ATA,GTA | I,V 3 | XP_011513355.1 |
ZPBP - zona pellucida binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |