Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGTGCTCAACCTGCTGCAGATC[C/G]AGAGGAATGTCACTCTCTTCCCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSIG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INSIG1 - insulin induced gene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005542.4 | 533 | Missense Mutation | CAG,GAG | Q,E 108 | NP_005533.2 | |
NM_198336.2 | 533 | Intron | NP_938150.2 | |||
NM_198337.2 | 533 | Missense Mutation | CAG,GAG | Q,E 108 | NP_938151.1 | |
XM_005249542.1 | 533 | Missense Mutation | CAG,GAG | Q,E 108 | XP_005249599.1 | |
XM_005249543.1 | 533 | Missense Mutation | CAG,GAG | Q,E 108 | XP_005249600.1 | |
XM_017012177.1 | 533 | Missense Mutation | CAG,GAG | Q,E 108 | XP_016867666.1 |