Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGAGGGAATATGGAGCAATCCC[A/G]GTCACAGCAGCGTGGGGGTGAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600639 MIM: 118425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP2 - caspase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLCN1 - chloride voltage-gated channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000083.2 | 101 | Missense Mutation | CAG,CGG | Q,R 5 | NP_000074.2 | |
XM_011515782.2 | 101 | Intron | XP_011514084.1 | |||
XM_017011739.1 | 101 | Intron | XP_016867228.1 | |||
XM_017011740.1 | 101 | Intron | XP_016867229.1 |