Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGGTCTGTCCCCACAGGCTGCAC[A/G]GGAGAAGGATGGTATCGTAATAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZKSCAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZKSCAN1 - zinc finger with KRAB and SCAN domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287054.1 | 296 | UTR 5 | NP_001273983.1 | |||
NM_001287055.1 | 296 | Intron | NP_001273984.1 | |||
NM_003439.2 | 296 | Missense Mutation | CAG,CGG | Q,R 18 | NP_003430.1 | |
XM_005250565.4 | 296 | Missense Mutation | CAG,CGG | Q,R 18 | XP_005250622.1 | |
XM_011516559.2 | 296 | Missense Mutation | CAG,CGG | Q,R 18 | XP_011514861.1 |