Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGCAACCTTTCTTTATGCCGAA[A/G]TAGGACTCATTTTAATCTTCTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAP29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAP29 - B-cell receptor-associated protein 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008405.2 | 381 | Missense Mutation | ATA,GTA | I,V 16 | NP_001008405.1 | |
NM_018844.3 | 381 | Missense Mutation | ATA,GTA | I,V 16 | NP_061332.2 | |
XM_006716051.2 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_006716114.1 | |
XM_011516406.2 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_011514708.1 | |
XM_011516407.1 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_011514709.1 | |
XM_011516408.2 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_011514710.1 | |
XM_011516409.2 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_011514711.1 | |
XM_017012436.1 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_016867925.1 | |
XM_017012437.1 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_016867926.1 | |
XM_017012438.1 | 381 | Missense Mutation | ATA,GTA | I,V 16 | XP_016867927.1 |
DUS4L - dihydrouridine synthase 4 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |