Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGATACTTTTAAATCACCTTT[C/G]AACTTTTCCACAGTAACCGTAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CDCA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDCA2 - cell division cycle associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317906.1 | 708 | Missense Mutation | TTC,TTG | F,L 40 | NP_001304835.1 | |
NM_001317907.1 | 708 | Missense Mutation | TTC,TTG | F,L 55 | NP_001304836.1 | |
NM_152562.3 | 708 | Missense Mutation | TTC,TTG | F,L 55 | NP_689775.2 | |
XM_006716294.2 | 708 | Missense Mutation | TTC,TTG | F,L 55 | XP_006716357.1 | |
XM_011544415.1 | 708 | Missense Mutation | TTC,TTG | F,L 55 | XP_011542717.1 | |
XM_011544416.2 | 708 | Missense Mutation | TTC,TTG | F,L 55 | XP_011542718.1 | |
XM_011544417.2 | 708 | Missense Mutation | TTC,TTG | F,L 55 | XP_011542719.1 | |
XM_011544418.1 | 708 | Missense Mutation | TTC,TTG | F,L 55 | XP_011542720.1 |
KCTD9 - potassium channel tetramerization domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |